Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD34B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734435 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4435 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4435 Supplier Proteintech Group Inc Supplier No. 245811AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ANKRD34B, also named as Ankyrin repeat domain-containing protein 34B, is a 514 amino acid protein, which contains 4 ANK repeats and belongs to the ANKRD34 family. The calculated molecular weight of ANKRD34B is 55 kDa, but the phosphorylated protein is about 60-65 kDa.

Specifications

Antigen ANKRD34B
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ANKRD34B
Gene Accession No. A5PLL1, Q3UUF8
Gene Alias ANKRD34B, ankyrin repeat domain 34B, DP58
Gene Symbols ANKRD34B
Host Species Rabbit
Immunogen ANKRD34B Fusion Protein Ag20346
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218440, 340120
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.