Learn More
Invitrogen™ Ankyrin 1 Monoclonal Antibody (N388A/10), APC

Description
A 1:100 dilution of MA5-45729 was sufficient for detection of Ankyrin R in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 200kDa. Does not cross-react with Ankyrin-B or Ankyrin-G. This antibody was formerly sold as clone S388A-10.
Specifications
Specifications
| Antigen | Ankyrin 1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N388A/10 |
| Concentration | 1 mg/mL |
| Conjugate | APC |
| Formulation | 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 |
| Gene | ANK1 |
| Gene Accession No. | P16157, Q02357 |
| Gene Alias | ANK; ANK1; ANK-1; ANK-1; erythrocyte skeletal protein; ankyrin 1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin-1; Ankyrin-R; erythrocyte ankyrin; nb; normoblastic anemia; pale; rCG_43073; SPH1; SPH2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.