Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ankyrin G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143596 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143596 100 μL
PIPA5143498 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143596 Supplier Invitrogen™ Supplier No. PA5143596
  • $581.00
    $494.65 / Each of 1

    Save $86.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Ankyrin G Polyclonal Antibody for ICC/IF, Western Blot

Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin G
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene ANK3
Gene Accession No. G5E8K5, O70511, Q12955
Gene Alias 2900054D09Rik; AI314020; ANK3; ANK-3; AnkG; ankyrin 3; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin G; ankyrin-3; Ankyrin-G; brain-specific ankyrin-G; LOW QUALITY PROTEIN: ankyrin-3; MRT37
Gene Symbols ANK3
Host Species Rabbit
Immunogen The C-terminal 398 amino acids of human ankyrin 3 expressed in and purified from E. coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11735, 288, 361833
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.