Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKZF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530665 Supplier Invitrogen™ Supplier No. PA530665
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (84%), Rat (86%), Chimpanzee (99%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays a role in the cellular response to hydrogen peroxide and in the maintenance of mitochondrial integrity under conditions of cellular stress (PubMed:28302725). Involved in the endoplasmic reticulum (ER)-associated degradation (ERAD) pathway. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKZF1
Applications Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ANKZF1
Gene Accession No. Q9H8Y5
Gene Alias 1300008P06Rik; 2810025E10Rik; ankyrin repeat and zinc finger domain containing 1; ankyrin repeat and zinc finger domain-containing protein 1; ankyrin repeat containing protein RGD1359242; ANKZF1; D1Ertd161e; RGD1359242; Zinc finger protein 744; ZNF744
Gene Symbols ANKZF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKZF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.