Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKZF1/ZNF744 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2266 Supplier Bioss Supplier No. BS12476R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFR is critical for Staufen 2 isoform specific nucleocytoplasmic shuttling in neurons.

Specifications

Antigen ANKZF1/ZNF744
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKZF1
Gene Accession No. Q9H8Y5
Gene Alias ankyrin repeat and zinc finger domain containing 1; Ankyrin repeat and zinc finger domain-containing protein 1; ANKZF1; Zinc finger protein 744; ZNF744
Gene Symbols ANKZF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKZF1/ZNF744.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55139
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.