Learn More
Foxx Life Sciences VersaCap 53mm (53B) Bottle and Flask Caps, Open Cap with Tangle Free Hose Barb Adapter, Two 1/8 Inch Barbs for 1/8 Inch ID Tubing
Supplier:  Foxx Life Sciences 2050317RLS
Encompass_Preferred
205-0317-RLS: VersaCap, 53mm, with 2x 1/8" Molded In HB Adapter. Patented VersaCap is leak-proof, uses a replaceable Pt-cured silicone gasket, and provides the ultimate in top-connection flexibility. Open VersaCap is compatible with a variety of adapter inserts, enabling users to quickly change the top connections of each carboy or bottle to match their application needs. VersaCap offers large finger grips that allow for easy tightening and opening. They are also stackable, and offered in 53mm, 83mm (compatible with most carboys), and 120mm wide-mouth versions that fit exclusively on Foxx Life Sciences 20L, 40L, and 75L EZfil carboys. Quick-connect hose barb options include combinations of two, three, and four ports, with 1/4 and 3/8 hose barb sizes. Molded-in hose barb options are available in 1/8, 1/4, 516, and 1/2 sizes. VersaCap are made from polypropylene, which may be autoclaved at 15 PSI and 121C (250F) for 20 minutes.
Catalog No. 50-104-9814
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.