Learn More
Invitrogen™ Annexin A10 Monoclonal Antibody (1F9)

Description
Peptide Sequence: MFCGDYVQGT IFPAPNFNPI MDAQMLGGAL QGFDCDKDML INILTQRCNA QRMMIAEAYQ SMYGRDLIGD MREQLSDHFK DVMAGLMYPP PLYDAHELWH AMKGVGTDEN CLIEILASRT NGEIFQMREA YCLQYSNNLQ EDIYSETSGH FRDTLMNLVQ GTREEGYTDP AMAAQDAMVL WEACQQKTGE HKTMLQMILC NKSYQQLRLV FQEFQNISGQ DMVDAINECY DGYFQELLVA IVLCVRDKPA YFAYRLYSAI HDFGFHNKTV IRILIARSEI DLLTIRKRYK ERYGKSLFHD IRNFASGHYK KALLAICAGD AEDY.
Specifications
Specifications
| Antigen | Annexin A10 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1F9 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ANXA10 |
| Gene Accession No. | Q9UJ72 |
| Gene Alias | annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.