Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A11 Monoclonal Antibody (24GB6225)
GREENER_CHOICE

Catalog No. MA557335
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA557335 100 μL
MA557336 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA557335 Supplier Invitrogen™ Supplier No. MA557335
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A11 Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A11
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 24GB6225
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ANXA11
Gene Accession No. P50995, P97384
Gene Alias 56 kDa autoantigen; A830099O17Rik; annexin; annexin A11; Annexin A11 (Annexin XI) (Calcyclin-associated annexin 50) (CAP-50); annexin XI; Annexin-11; ANX11; Anxa11; autoantigen; autoantigen, 56-kD; calcyclin-associated annexin 50; Calcyclin-associated annexin-50; CAP50; CAP-50; OTTHUMP00000019957; OTTHUMP00000019958; RP11-369J21
Gene Symbols ANXA11
Host Species Mouse
Immunogen Recombinant protein of human ANXA11 (aa 1-505).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11744, 290527, 311
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.