Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Annexin A11 Protein
SDP

Catalog No. NB170777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170777 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170777 Supplier Novus Biologicals™ Supplier No. NBP150857250UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-505 of Human Annexin A11 The Recombinant Human Annexin A11 Protein is derived from E. coli. The Recombinant Human Annexin A11 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS containing 20% glycerol, 2 mM DTT, 100 mM NaCl
Molecular Weight (g/mol) 56.6 kDa
Purification Method SDS-PAGE
Quantity 250 μg
Source E. coli
Immunogen ANXA11, 1-505 aa, Human, His tag, E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Annexin A11
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.