Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A2 Monoclonal Antibody (Z014)
GREENER_CHOICE

Catalog No. 034400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-440-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-440-0 Supplier Invitrogen™ Supplier No. 034400
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Annexin A2 Monoclonal Antibody for Western Blot, IHC, ICC/IF, ELISA

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Z014
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation Proprietary buffer with 0.09% sodium azide
Gene ANXA2
Gene Accession No. A2SW69, P04272, P07355, Q6TEQ7
Gene Alias 33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I
Gene Symbols ANXA2
Host Species Mouse
Immunogen Annexin II purified from bovine lung.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100048993, 282689, 302, 403435
Target Species Bovine, Canine, Human, Sheep
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.