Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin A2 Recombinant Rabbit Monoclonal Antibody (K01_1M78)
GREENER_CHOICE

Catalog No. MA563294
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563294 100 μL
MA564753 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563294 Supplier Invitrogen™ Supplier No. MA563294
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: FDAERDALNI ETAI.

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1M78
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ANXA2
Gene Accession No. P07355, P07356, Q07936
Gene Alias 33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I
Gene Symbols ANXA2
Host Species Rabbit
Immunogen A synthetic peptide of human Annexin A2 (aa 33-46).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12306, 302, 56611
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.