Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Annexin A3, Mouse, Unlabeled, Clone: 9996-1-3M1/3L7-5,

Catalog No. MABS1329 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1329 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1329 Supplier MilliporeSigma™ Supplier No. MABS1329
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Annexin A3 (UniProt P12429; also known as 35-alpha calcimedin, Annexin III, Annexin-3, Inositol 1,2-cyclic phosphate 2-phosphohydrolase, Lipocortin III, PAP-III, Placental anticoagulant protein III) is encoded by the ANXA3 (also known as ANX3) gene (Gene ID 306) in human. Annexin A3 is a Ca2+-dependent phospholipid-binding protein shown to promote angiogenesis and rat liver regeneration. Upregulated annexin A3 expression is reported in ovarian, breast, colon, lung, gastric, gallbladder, testicular, and urothelial cancers, where it is involved in promoting tumorigenesis and resistance to chemotherapy. ANXA3 is reported to represent the most significantly upregulated gene that encodes for a secretory protein in the CD133+ liver cancer stem cell (CSC) subset. Both cellular and secretory annexin A3 play pivotal roles in promoting cancer and stem cell-like features in CD133+ liver CSCs through a dysregulated JNK pathway. Annexin A3 serum levels in hepatocellular carcinoma (HCC) patients are closely associated with aggressive clinical features. Annexin A3 neutralizing antibody is shown to cause a significant reduction in HCC tumor growth and self-renewal by effectively killing off CD133+ CSCs both in cultures in vitro and in animal xenograft in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A3
Applications Neutralization, Western Blot
Classification Monoclonal
Clone 9996-1-3M1/3L7-5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in Tris-buffered saline (TBS) containing 50mM tris-HCl (pH7.5) and 150mM NaCl without preservatives.
Gene ANXA3, ANX3
Gene Accession No. P12429
Host Species Mouse
Immunogen Recombinant human annexin A3.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005130
Test Specificity This monoclonal antibody detected a single ∽33 kDa target band among a panel of human hepatocellular carcinoma (HCC) cell lines. The epitope has been mapped to a 10-a.a. region within the third annexin domain (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.