Learn More
Description
- Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells.
- Annexin V staining, paired with 7-AAD or PI, is widely used to identify apoptotic stages by flow cytometry
Specifications
Specifications
| Conjugate | Biotin |
| Product Type | Apoptosis Kit |
| For Use With (Application) | Flow Cytometry |
| Kit Contents | Annexin V-Biotin protein, Binding Buffer (4X), Propidium Iodide (20μg/mL) |
| Quantity | 100 Tests |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
