Learn More
Description
- Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells.
- Annexin V staining, paired with 7-AAD or PI, is widely used to identify apoptotic stages by flow cytometry
Specifications
Specifications
| For Use With (Application) | Flow (Cell Surface), Immunocytochemistry/Immunofluorescence |
| Product Type | Apoptosis Kit |
| Content And Storage | Annexin V-FITC Protein, Propidium Iodide, 10X PBS, 10X Binding Buffer, Control Cells: 1x106 cells/mL. Store at 4°C. Do not freeze. |
| Conjugate | FITC |
| Product Line | Novus |
| Quantity | 100 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
