Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Annexin V Apoptosis Kit, Fluorescein
SDP

Catalog No. NB170650 Shop All Novus Products
Change view
Click to view available options
Quantity:
125 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170650 -
NB483001K 125 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170650 Supplier Novus Biologicals™ Supplier No. NB483001K250TESTS

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Allows for distinction between early apoptotic and viable cells.

  • Annexin V is a member of a calcium and phospholipid binding family of proteins with vascular anticoagulant activity. It is also known as PP4, PAP I, CPB-I, CaBP33, VACa, anchorin CII, lipocortin-V, endonexin II, and thromboplastin inhibitor
  • Facilitates rapid fluorometric quantitation of apoptotic cell
  • Allows the use of streptavidinconjugated fluorophores other than FITC for detection

Applications: Flow cytometry
Conjugate: Fluorescein
Sample type: Unfixed cells

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.