Learn More
Novus Biologicals™ Annexin V Apoptosis Kit, Fluorescein

Description
- Annexin V is a member of a calcium and phospholipid binding family of proteins with vascular anticoagulant activity. It is also known as PP4, PAP I, CPB-I, CaBP33, VACa, anchorin CII, lipocortin-V, endonexin II, and thromboplastin inhibitor
- Facilitates rapid fluorometric quantitation of apoptotic cell
- Allows the use of streptavidinconjugated fluorophores other than FITC for detection
Applications: Flow cytometry
Conjugate: Fluorescein
Sample type: Unfixed cells
Specifications
Specifications
| Quantity | 125 Tests |
| Product Type | Apoptosis detection kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Protect from light. Do not use past expiration date. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.