Learn More
AdipoGen Annexin V (human) (rec.) (FITC) (20 tests)
SDP
Supplier:  AdipoGen AG40B0005FT020
Encompass_Preferred
Protein. Recombinant human annexin V fused to a GST-tag. Source: E. coli. Liquid. In PBS, pH 7.4, containing 0.02% sodium azide. Binds to phosphatidylserine (PS). Purity: >95% (SDS-PAGE). Annexin V is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade. Annexin V competes for phosphatidylserine binding sites (e.g. platelets) with prothrombin or coagulation factors and inhibis the activity of phospholipase A1. Antibodies directed against annexin V are found in patients with a disease called the antiphospholipid syndrome(APS), a thrombophilic disease associated with autoantibodies against phospholipid compounds. Annexin V is used as a probe in the "annexin V affinity assay" to detect cells that have expressed phosphatidylserine on the cell surface, a feature found in apoptosis and other forms of cell death.
Catalog No. 50-205-3070
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.