Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Annexin V Ready Flow Conjugates for Apoptosis Detection

Catalog No. R37174
Encompass_Preferred
Change view
Click to view available options
Excitation/Emission:
410/455
495/519
650/660
650/665
Color:
Blue
Far-Red
Green
Red
Conjugate:
APC (Allophycocyanin)
Alexa Fluor 488
Alexa Fluor 647
Pacific Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Excitation/Emission Color Conjugate
R37174 495/519 Green Alexa Fluor 488
R37175 650/665 Far-Red Alexa Fluor 647
R37176 650/660 Red APC (Allophycocyanin)
R37177 410/455 Blue Pacific Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R37174 Supplier Invitrogen™ Supplier No. R37174
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Annexin V conjugate stains enable you to easily determine apoptotic cells via flow cytometry.

Enjoy the convenience of ready-to-use Annexin V Ready Flow stand-alone conjugate stains, which offer up to 100-fold fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry. There is no need to dilute, weigh, or pipette the conjugate stains; simply add one drop into cells, incubate, and analyze.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

The Annexin V Ready Flow conjugates are provided as ready-to-use, direct-to-sample apoptotic cell stains. Simply add one drop from the dropper bottle directly into 1 x 105 cells in 2.5 mM Ca2+-containing cell culture medium or annexin binding buffer (Cat. Nos. V13246 or BMS500BB), incubate, and analyze. The traditional annexin staining protocol can be used as an alternative method by replacing the addition of 5 μL annexin conjugate with the addition of 1 drop from the Ready Flow dropper bottle, following the volumes and incubation conditions as indicated.

The Annexin V Alexa Fluor 488 conjugate is excitable at 495 nm with an emission peak at 519 nm.

The Annexin V Alexa Fluor 647 conjugate is excitable at 650 nm with an emission peak at 665 nm.

The Annexin V APC conjugate is excitable at 650 nm with an emission peak at 660 nm.

The Annexin V Pacific Blue conjugate is excitable at 410 nm with an emission peak at 455 nm.

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

TRUSTED_SUSTAINABILITY

Specifications

Description Annexin V Alexa Fluor 488 Ready Flow Conjugate
Quantity 2 Dropper Bottles
Product Type Ready Flow Conjugate
Content And Storage Store at 2-8°C.
Flow Cytometer Laser Lines Blue
Excitation/Emission 495/519
No. of Reactions 120
Color Green
Shipping Condition Room Temperature
Conjugate Alexa Fluor 488
For Use With (Equipment) Flow Cytometer
Show More Show Less
I would like to label only 100 µL of sample of the same cell density using a Ready Flow product. The manual notes 2 drops per 1 mL (1 X 10E6 cells/mL). May I scale down for smaller volumes?

Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.

What is the concentration of dye/reagent in the Ready Flow products?

The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.