Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO1 Mouse anti-Human, Clone: UMAB250, liquid, UltraMAB™
SDP

Catalog No. 501685590
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5590 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5590 Supplier OriGene Supplier No. UM870142
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.

Specifications

Antigen ANO1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB250
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ANO1
Gene Accession No. Q5XXA6
Gene Alias DOG1, ORAOV2, TAOS2, TMEM16A
Gene Symbols ANO1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-333 of human ANO1 (Dog1) produced in E.coli.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.