Learn More
Description
Recommended positive controls: mouse heart, rat heart. Predicted reactivity: Mouse (80%), Rat (80%), Dog (85%), Pig (87%), Rabbit (81%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ANP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.44 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Gene | NPPA |
| Gene Accession No. | P01160, P01161, P05125 |
| Gene Alias | Anf; ANP; ATFB6; atrial natriuretic factor; Atrial natriuretic peptide; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; ATRST2; Auriculin-A; Auriculin-B; Cardiodilatin; Cardiodilatin-related peptide; cardionatrin; CDD; CDD-ANF; CDP; natriuretic peptide A; Natriuretic peptide precursor A (pronatriodilatin, also Anf, Pnd); natriuretic peptide precursor A variant 1; natriuretic peptide precursor A variant NPPA-M1; natriuretic peptide precursor A variant NPPA-M2; natriuretic peptide precursor A variant NPPA-M3; natriuretic peptide precursor type A; natriuretic peptide type A; natriuretic peptides A; Nppa; Pnd; Prepronatriodilatin; Pronatriodilatin; RATANF; urodilatin |
| Gene Symbols | NPPA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
