Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANP Recombinant Rabbit Monoclonal Antibody (JM08-36)
GREENER_CHOICE

Catalog No. PIMA532774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532774 Supplier Invitrogen™ Supplier No. MA532774
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP
Applications Western Blot
Classification Recombinant Monoclonal
Clone JM08-36
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene NPPA
Gene Accession No. P01160
Gene Alias Anf; ANP; ATFB6; atrial natriuretic factor; Atrial natriuretic peptide; atriopeptin; Atriopeptin I; Atriopeptin II; Atriopeptin III; Atriopeptin-1; Atriopeptin-2; Atriopeptin-3; ATRST2; Auriculin-A; Auriculin-B; Cardiodilatin; Cardiodilatin-related peptide; cardionatrin; CDD; CDD-ANF; CDP; natriuretic peptide A; Natriuretic peptide precursor A (pronatriodilatin, also Anf, Pnd); natriuretic peptide precursor A variant 1; natriuretic peptide precursor A variant NPPA-M1; natriuretic peptide precursor A variant NPPA-M2; natriuretic peptide precursor A variant NPPA-M3; natriuretic peptide precursor type A; natriuretic peptide type A; natriuretic peptides A; Nppa; Pnd; Prepronatriodilatin; Pronatriodilatin; RATANF; urodilatin
Gene Symbols NPPA
Host Species Rabbit
Immunogen Synthetic peptide within Human ANP aa 102-151.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4878
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.