Learn More
Invitrogen™ ANP32B Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DNCKSNDWKI ESLTAEFVNL EFLRLI Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 81%.
Specifications
Specifications
| Antigen | ANP32B |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ANP32B |
| Gene Accession No. | Q92688 |
| Gene Alias | 2410015B15Rik; acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; acidic leucine-rich nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family, member B; acidic protein rich in leucines; Anp32b; APRIL; OTTHUMP00000063770; Pal31; PHAPI2; PHAPI2a; proliferation related acidic leucine rich protein PAL31; proliferation-related acidic leucine-rich protein PAL31; Putative HLA-DR-associated protein I-2; silver-stainable protein SSP29; SSP29; TNF superfamily member 13 |
| Gene Symbols | ANP32B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.