Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANP32D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575586 Supplier Invitrogen™ Supplier No. PA575586
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ANP32D (acidic (leucine-rich) nuclear phosphoprotein 32 family, member D), also known as PP32R2 (phosphoprotein 32-related protein 2) or tumorigenic protein pp32r2, is a 131 amino acid protein that contains 3 leucine-rich repeats and belongs to the ANP32 family. ANP32D shares 89.3% amino acid sequence identity with ANP32A, a tumor suppressor that inhibits a variety of cancers, such as prostate and breast cancers. Although similar to ANP32A, ANP32D is tumorigenic and is preferentially expressed in prostatic cancer. Conversely, it is uncertain that ANP32D is neurally expressed. Intronless, ANP32D may be a pseudogene that is expressed in pathological situations and may also modulate the oncogenic potential of human prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANP32D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ANP32D
Gene Accession No. O95626
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member D; acidic leucine-rich nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32 family member D; acidic nuclear phosphoprotein 32D; ANP32D; Phosphoprotein 32-related protein 2; pp32 related 2; PP32R2; Tumorigenic protein pp32r2
Gene Symbols ANP32D
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human ANP32D.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23519
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.