Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Sterile gloves post-cleaned and packed in a bulk poly bag with an additional sterilization step
- Manufactured in a facility registered to ISO 9001, ISO 13485, ISO14001
- High puncture resistance three times more than standard latex gloves
- Protects against a wide range of chemicals
- Free of natural rubber latex proteins
- Textured fingertips
- Rolled beaded cuff, Ambidextrous, Powder free
Specifications
Specifications
| Certifications/Compliance | ASTM D6978-05, FDA 21 CFR 177.2600 |
| Certifications/Compliance (Europe) | Category III, EAC TP TC 019:2011, EN 420:2003 + A1:2009, EN ISO 374-1:2016 |
| Cleanroom Class | Class 100 (ISO 5) |
| Color | Green |
| Exterior Finish | Partially Textured |
| Finger Thickness (English) | 6.3 mil |
| Finger Thickness (Metric) | 0.16 mm |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Nitrile |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?


