Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell TouchNTuff™ 93-700 Sterile Nitrile Cleanroom Gloves
BIOPHARMA

Catalog No. 19823521 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-521 X-Large
19-823-518 Small
19-823-519 Medium
19-823-520 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-823-521 Supplier Ansell Supplier No. 384233
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile nitrile gloves deliver protection and durability when transferring liquids. Compatible with Class 100 (ISO 5) environments.

  • Sterile gloves post-cleaned and packed in a bulk poly bag with an additional sterilization step
  • Manufactured in a facility registered to ISO 9001, ISO 13485, ISO14001
  • High puncture resistance three times more than standard latex gloves
  • Protects against a wide range of chemicals
  • Free of natural rubber latex proteins
  • Textured fingertips
  • Rolled beaded cuff, Ambidextrous, Powder free
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6978-05, FDA 21 CFR 177.2600
Certifications/Compliance (Europe) Category III, EAC TP TC 019:2011, EN 420:2003 + A1:2009, EN ISO 374-1:2016
Cleanroom Class Class 100 (ISO 5)
AQL 1.5
Color Green
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.1 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size X-Large
Sterility Sterile
Thickness (English) 6.3 mil
Thickness (Metric) 0.16 mm
EN 421 Yes
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Manufacturing/QMS Audit Standards ISO 9001:2015, ISO 13485, ISO 14001
Model 384233
Series TouchNTuff™ 93-700
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.