Learn More
Ansell™ Chemical- and Flame-Resistant Bib Pants
Catalog No. 17987444F Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Red
Size:
Large
X-Large
2X-Large
Size (English) Waist:
52 in.
56 in.
60 in.
Material:
CPC Polyester
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Size (English) Waist Material
17-987-444F Blue 2X-Large 60 in. -
17-987-449F Red 2X-Large 60 in. CPC Polyester
17-987-449E Red X-Large 56 in. CPC Polyester
17-987-449D Red Large 52 in. CPC Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.
Catalog No. 17-987-444F Supplier Ansell™ Supplier No. 666722XL
  • $2,950.00
    $2,360.65 / Each of 1
    Save $589.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gore™ ChemSplash™ fabric is vapor permeable and should not be used for protection against hazardous vapors, carcinogens, or other life-threatening materials. Nomex™ and Gore ChemSplash fabric are not always in stock. Call your Fisher Safety Customer Service Representative for current information.
Lightweight and breathable CPC garments made with Gore™ ChemSplash™ fabric reduce overheating and fatigue while preventing liquid chemical splash penetration
Gore membrane is a bicomponent PTFE membrane that prevents liquid penetration but allows a vapor permeation, allowing perspiration to evaporate and cool the wearer.
  • Garments stitched with Gore thread
  • All seams are heat and pressure sealed with Gore-Seam™ tape to prevent liquid penetration
  • Fabrics are durable, flexible, tear and snag resistant, washable, and reusable
Coveralls
  • Feature two-way, heavy-duty, static-free zipper
  • Double storm flap; stand-up collar; raglan sleeves
  • Hook-and-loop closures on storm flap, collar, sleeves, and leg bottoms
Jackets
  • Heavy-duty zipper
  • Double storm flap; stand-up collar; raglan sleeves
  • Hook-and-loop closures on storm flap, collar, and sleeves
  • Sized to be worn over clothing
Bib Pants
  • Elastic suspenders with quick-release clips for fast removal
  • Leg bottoms have hook-and-loop closures
Regular Hoods
  • Attach, by means of a hook-and-loop fastener, to back of collar on coveralls or jacket
  • Drawstring closure gives snug fit over hardhat
Order Info
For chemical splash protection, choose CPC garments made with red polyester Gore ChemSplash fabric (7 oz. per sq. yd.). When flame protection is also required, choose garments made with blue aramid (Nomex™) Gore ChemSplash fabric (6 oz. per sq. yd.); when exposed to flame, it will self-extinguish when removed from the flame source, and not contribute to burn injury.
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Double Front Flap with Adjustable Hook and Loop Straps
Color Blue
Size 2X-Large
Suspender Elastic
Construction Trilaminate
For Use With (Application) Industries, bulk chemical handling and transport, chemical plants and transportation, chemical processing, Oil refining and petrochemicals, handling specialty chemicals
Length (English) Inseam 31 in.
Length (Metric) Inseam 79 cm
Size (English) Waist 60 in.
Certifications/Compliance NFPA 70E™, ASTM F 1891-02 ASTM
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.