Learn More
Ansell™ CPP White Vinyl Aprons
Catalog No. 19058830 Shop All Ansell Products
Change view
Click to view available options
Dimensions (L x W):
35 x 45 in.
Catalog No. Dimensions (L x W)
19-058830 35 x 45 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-058830 Supplier Ansell™ Supplier No. 950102
  • $875.00
    $375.65 / Case of 12
    Save $499.35
    57% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heavy-duty protection in a variety of styles
Highest-quality virgin vinyl in 4, 6, 8, 12 and 18mil thicknesses
Benefits:
  • Superior resistance to chemicals, fats, oils, greases, punctures and abrasions
  • Optional features:
    • Stomach patches for extended wear
    • Raw or sewn edges
    • Three-piece ties
    • Rough texture
    Food Preparation, Processing and Handling:
    • Beef/pork/poultry/fish processing
    • Dairy processing
    Public Utilities:
    • Cleaning and de-greasing
    • Industrial maintenance
    • Janitorial and maintenance work
    Chemical:
    • Chemical processing, oil refining and petrochemicals
    TRUSTED_SUSTAINABILITY
    Specifications
    Material PVC
    Color White
    Thickness 18 mil
    Dimensions (L x W) 35 x 45 in.
    Adhere to
    Safety Regulations and Best Practices
    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.