Learn More
Ansell Dura-Touch™ Powder Free Stretch Vinyl Gloves
Catalog No. 19162024 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-024 X-Large
19-162-021 Small
19-162-022 Medium
19-162-023 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-162-024 Supplier Ansell Supplier No. 34175 XL 105016
This item is not returnable. View return policy
Latex comfort without the risk of latex allergies
  • Made of PVC for comfort and dexterity
  • Beaded cuff
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Vinyl
Size X-Large
Thickness (English) 5 mil