Learn More
Description
- Proprietary formulation delivers a soft and sensitive yet robust surgical glove
- Anti-slip, smooth external surface treatment for instrument handling and grip
- DERMASHIELD Technology™ internal coating
- Tested for use with chemotherapy drugs
- Extra comfort during long, sustained procedures
- Damp donning
- Use as an outer glove when double gloving
Specifications
Specifications
| Certifications/Compliance | ASTM D3577, ASTM D6978-05 |
| Certifications/Compliance (Europe) | EN 16523-1, EN 374-2 and -4, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10282 |
| Accelerator Free | No |
| AQL | 0.65 |
| Color | Natural |
| Disposable | Single-use |
| Exterior Finish | No Texture |
| Hand | Specific |
| Laboratory/Cleanroom Use | No |
| Latex Free | No |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

