Learn More
Ansell Encore Acclaim Latex Sterile Gloves
GREENER_CHOICE
Catalog No. 19166676 Shop All Ansell Products
Change view
Click to view available options
Color:
Cream
Catalog No. Color
19-166-676 Cream
1 options
This item is not returnable. View return policy
Catalog No. 19-166-676 Supplier Ansell Supplier No. 5795001
  • $1,018.00
    $699.65 / Case of 200
    Save $318.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex, powder free surgical glove with a comfortable, natural fit and feel.
Featuring Ansell formers and glove formulation, this glove's combination of fit and comfort together with optimized tensile strength, puncture and tear resistance properties enables healthcare workers to perform at their very best.
  • Proprietary formulation delivers a soft and sensitive yet robust surgical glove
  • Unique anti-slip, smooth external surface treatment for exceptional instrument handling and grip
  • DERMASHIELD Technology™ internal coating facilitates dry and damp donning as well as intra-operative changes
  • Tested for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D 1076, ISO9001, ISO13485, ASTM D 3577, Type I
Accelerator Free No
AQL 0.65
Color Cream
Disposable Single-use
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Latex
Powdered Powder Free
Product Type Surgical Gloves
Size 6
Sterility Sterile
Thickness (English) 9 mil
Length (English) 11.7 in.
Length (Metric) 29.7 cm