Description
- 100% Kevlar™ aramid fiber material provides heavy cut protection
- Protects from sharp edges of metals, ceramics, glass and other materials
- Reversible design allows glove to be worn on either hand
- Launderable
Specifications
Specifications
| Coating Material | Uncoated |
| Color | Yellow |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Medium |