Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 27-602 Fully Nitrile Coated Heavy Duty Cotton Jersey Gloves

Catalog No. 11391127B Shop All Ansell Products
Click to view available options
Size:
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Ansell's unique nitrile coating formulation offers resistance to sharp/abrasive materials, oil, and grease. The heavy duty design is durable, and certified clean for metals and motor manufacturing.

  • Ansell's unique nitrile formulation offers resistance to sharp and abrasive materials
  • Gloves provides a barrier against grease and oil that will not degrade
  • Guaranteed clean for metals and motor manufacturing
  • Three times more durable than heavy-duty leather
  • Stronger and more flexible than PVC
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 27-602
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.