Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated

Catalog No. 1913011898F Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-11898F 11
19-130-11898A 6
19-130-11898B 7
19-130-11898C 8
19-130-11898D 9
19-130-11898E 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-11898F Supplier Ansell Supplier No. 205679
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ergonomic design reduces hand fatigue and provides a fit that mimics bare hands. Nitrile foam coating enhances tactile sensitivity and grip capabilities in dry or light oil conditions.

  • Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
  • Aramid fiber provides an additional level of protection
  • Ergonomic design provides bare-hand closeness and reduces hand fatigue
  • Nitrile foam coating enhances grip capabilities and tactile sensitivity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Series 11-501
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.