Learn More
Ansell HyFlex™ 11-920 Nitrile Coated Oil Repellent Gloves
Catalog No. 19148502 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-502 Medium
19-148-501 Small
19-148-504 X-Large
19-148-503 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-148-502 Supplier Ansell Supplier No. 255003
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Seamless knit gloves feature ANSELL GRIP™ Technology which utilizes microscopic channels in the glove coating to displace oil, enhancing grip capabilities when working with oily parts.
  • High performance grip and oil repellence when working in oily environments
  • ANSELL GRIP™ Technology contains microscopic channels in the glove coating to displace oil
  • RIPEL™ Liquid Repellence Technology creates a liquid impermeable layer to prevent oil or lubricant from making contact with the wearer's skin
  • Seamless knit design provides a comfortable fit with maximum dexterity
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Size Medium
For Use With (Application) Automotive Assembly, Equipment Operation, Machinery, and Metal Fabrication
Glove Cuff Style Knit
Series 11-920
Yarn Gauge 15
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.