Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-100 Self-Sanitizing Industrial Gloves

Catalog No. 19046720A Shop All Ansell Products
Change view
Click to view available options
ANSI Abrasion Rating:
ANSI 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ANSI Abrasion Rating
19-046-720A ANSI 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-046-720A Supplier Ansell Supplier No. 205589
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed with Ionic+™, an EPA-registered anti-microbial technology that leverages the anti-microbial power of silver to protect gloves from microbial impacts.

  • Designed with Ionic+™, an EPA-registered antimicrobial technology (EPA Registration #70927-6)
  • Leverages the antimicrobial power of silver to protect gloves from microbial impacts
  • Protects glove from bacterial-related odor, stain, and fabric degradation
  • Self-Sanitizing 3600° antimicrobial protection of both the liner and coating
  • Touchscreen compatible for use with a wide range of devices
  • All day comfort with high dexterity in a light-weight breathable design
  • Coating will not inhibit microbial action
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ANSI/ISEA 105-2024 (Abrasion Level 3, Cut Level A1), EN 388:2016+A1:2018 (2131A), EN ISO 21420:2020, EU Biocides Regulation 528/2012, REACH
Certifications/Compliance (Europe) EU Biocides Regulation 528/2012
Chemotherapeutic Rated No
Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile Foam
Color Gray, White
Cut Resistance Rating ANSI A1
Disposable Reusable
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 6
EN388 Standard 2131A
For Use With (Application) Automotive Assembly, Hospitality, Logisitics, Transportation, Utilities, and Waste Disposal
Gender Unisex
Glove Cuff Style Knit
Lining Ionic+™ Active Antimicrobial
Series HyFlex™ 11-100
Yarn Gauge 15
Length (English) 8.03 to 10.78 in.
Length (Metric) 204 to 274 mm
Product Line HyFlex
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.