Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-818 Foam Nitrile Coated Nylon/Spandex Industrial Gloves

Catalog No. 19148413 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-413 9
19-148-411 7
19-148-412 8
19-148-414 10
19-148-415 11
19-148-410 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-148-413 Supplier Ansell Supplier No. 235139
Only null left
Add to Cart
Add to Cart

Ultralight multi-purpose gloves, designed for comfortable, tactile handling. Seamless, 18 gauge liner combined with an ultra-thin coating provides a bare hand feel for precision tasks that require fingertip sensitivity.

  • Soft, 18 gauge nylon/spandex liner provides a sheer, second skin for increased user comfort and compliance
  • Ultrathin FORTIX™ nitrile patent-pending 0.7 mm foam coating enhances dry grip and lasts 2x longer than traditional coatings
  • Coating is 20% more breathable that previous formulations for cooler, drier hands
  • ZONZ™ Comfort Fit Technology provides optimized knitting at the base of the pinky finger, across the finger tips, and through the palm
  • Benefits from proprietary washing process efficiently extracts impurities to deliver cleaner, more comfortable gloves
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Construction Knit
Coating Material Nitrile Foam
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish Rough
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Automotive Assembly, Machinery, Equipment Operation, and Chemical Handling
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series 11-818
Yarn Gauge 18
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.