Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 28-507 Nitrile Coated Aramid Fiber Gloves

Catalog No. 19048374 Shop All Ansell Products
Change view
Click to view available options
Size:
8
10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048374 8
19-048376 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-048374 Supplier Ansell Supplier No. 285813
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a two-piece aramid fiber liner for cut resistance in high stress areas. Nitrile safety cuff provides additional wrist and forearm protection.

  • Flexible nitrile coating
  • Provides cut and abrasion resistance
  • Enhanced grip capabilities
  • Ideal for handling metal sheets/panels, inspections, and shop operation
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Nitrile
Color Brown, Natural
Hand Specific
Material Aramid Fiber
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive Assembly, and Machine Operation
Glove Cuff Color Gold
Glove Cuff Style Safety
Series 28-507
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.