Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 28-507 Nitrile Coated Aramid Fiber Gloves

Catalog No. 19048376 Shop All Ansell Products
Change view
Click to view available options
Size:
8
10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048376 10
19-048374 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-048376 Supplier Ansell Supplier No. 285816
  • $412.91
    $208.65 / Dozen of 12

    Save $204.26

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,261.67
    $1,142.65 / Case of 6 DZ

    Save $1,119.02

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a two-piece aramid fiber liner for cut resistance in high stress areas. Nitrile safety cuff provides additional wrist and forearm protection.

  • Flexible nitrile coating
  • Provides cut and abrasion resistance
  • Enhanced grip capabilities
  • Ideal for handling metal sheets/panels, inspections, and shop operation
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105:2024 A2, EN 388:2016+A1:2018 4231B, EN ISO 21420:2020
Certifications/Compliance (Europe) EN 388:2016+A1:2018 4231B, EN ISO 21420:2020, REACH
Coated Area Palm
Coating Material Nitrile
Color Brown, Natural
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive Assembly, and Machine Operation
Glove Cuff Color Gold
Glove Cuff Style Safety
Resistant To Cut, Abrasion
Series 28-507
Length (English) 9.5 to 11.1 in.
Length (Metric) 242 to 282 mm
Product Line ActivArmr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.