Learn More
Ansell Neoprene Sleeve VSW-HS 59-050
Catalog No. 19053730 Shop All Ansell Products
Change view
Click to view available options
Cuff Style:
Elastic
Catalog No. Cuff Style
19-053730 Elastic
1 options
This item is not returnable. View return policy
N/A
Catalog No. 19-053730 Supplier Ansell Supplier No. 950235
  • $31.01
    $25.65 / Pair
    Save $5.36
    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,716.02
    $2,246.65 / Case of 96 PR
    Save $469.37
    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
N/A
Neoprene sleeve features elastic on both ends and is ideal for a wide range of chemical applications.
  • Double-insulating thermal liner: Resistance to both hot and cold
  • High-grade neoprene coating: Advanced mechanical and chemical resistance
  • Flexible, grip-enhancing design: Maneuverability and confident handling
TRUSTED_SUSTAINABILITY
Specifications
Size Standard
Sterility Non-sterile
Disposable Reusable
Closure Type Elastic Ends
Cuff Style Elastic
Finish Sealed
Latex Free No
Model 59-050
Thickness 16.9 mil
For Use With (Application) Chemical Processing, Electronics Manufacturing, Food Processing, Forestry, Oil Refining, Pharmaceutical
Length (Metric) 45.7 cm
Packaging 12 Pairs/Bag, 8 Bags/Cs.
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
N/A