Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 28-359 Nitrile Coated Heavyweight Gloves with NitraGold™ Cuff

Catalog No. 19058551 Shop All Ansell Products
Change view
Click to view available options
Size:
9
10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-058-551 10
19-058-550 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-058-551 Supplier Ansell Supplier No. 216150
  • $3,410.16
    $1,718.65 / Case of 72

    Save $1,691.51

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves combine the strength of aramid fibers with the grip and flexibility of Ansell's advanced nitrile coating technology. NitraGold&Trade; cuff provides mechanical protection against cuts and abrasion.

  • Heavyweight design provides a substantial level of protection
  • Aramid fiber construction is strong and durable
  • Nitrile coating is oil resistant and enhances grip capabilities
  • NitraGold™ cuff provides mechanical protection against cuts and abrasion
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Cuff Material NitraGold™
Coating Material Nitrile
Color Black, Gold
Cut Resistance Rating ANSI A3
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nitrile
Product Type Dip-coated Gloves
Size 10
EN388 Standard 4321C
For Use With (Application) Automotive Assembly/Repair
Liner Material Aramid Fiber
Series ActivArmr™ 28-359
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.