Learn More
Ansell Nitrilite™ Powder Free Nitrile Gloves
Catalog No. 19087910 Shop All Ansell Products
Change view
Click to view available options
Size: X-Large
Catalog No.
19-087910 Size: X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-087910 Supplier Ansell Supplier No. 93112XL
This item is not returnable. View return policy
Manufacturer-suggested Class M3.5 (Class 100) compatible
Designed specifically for Class M3.5 (Class 100) environments, these 100% nitrile powder free gloves are significantly low in particulate and extractable levels. Contain no plasticizers, silicone or pigments. Ideal for handling sensitive parts and materials. Stronger yet thinner (4 mil) than latex and vinyl, they're light and comfortable, with excellent dexterity and tactility.Ambidextrous. Color: natural.
TRUSTED_SUSTAINABILITY
Specifications
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Length (English) 12 in.
Length (Metric) 30.5 cm