Learn More
Ansell™ Sawyer-Tower™ 66-671 Series Coats
Catalog No. 19010041 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-041 X-Large
19-010-039 Medium
19-010-040 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-010-041 Supplier Ansell™ Supplier No. 66671XL
  • $2,919.00
    $1,815.65 / Case of 5
    Save $1,103.35
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Full-coverage chemical protection with added safety of fire resistance
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistance
  • HRC rating: 2
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material CPC Nomex Trilaminate
Color Blue
Arc Flash Rating 2
Resistance Flame, chemical
Length (English) 49 in.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.