Learn More
Ansell™ Sawyer-Tower™ 66-672 Series Bib Overalls
Catalog No. 19010045 Shop All Ansell Products
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
19-010-045 2X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-010-045 Supplier Ansell™ Supplier No. 666722XL/DEL
This item is not returnable. View return policy
Full-coverage chemical protection with added safety of fire resistance
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistance
  • HRC rating: 2
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material CPC Nomex Trilaminate
Size 2X-Large
Arc Flash Rating 2
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.