Learn More
Ansell™ Sol-Vex™ Nitrile Gloves
Resistant to chemicals—will not weaken or swell in many solvents, caustics or animal fats. Ansell™ Sol-Vex™ Nitrile Gloves are ideal for numerous applications involving chemical processing.
Manufacturer:  Ansell™ 37155 7 117141
 View more versions of this product
Catalog No. 11-391-137A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Straight
None
Contoured
15mil
Green
33cm
chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Nitrile
7
21 CFR 177-2600, FDA
13 in.