Learn More
Description
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
- Does not contain natural rubber latex proteins
- Strong grip in wet or dry conditions
- Rolled beaded for easy donning
- Powder free, Polymer-coated
- Ambidextrous
- Textured Fingers
- AQL: 1.5
Specifications
Specifications
| Chlorinated | Yes |
| Applications | Laboratory |
| AQL | 1.5 |
| Color | Green |
| Disposable | Single-use |
| Finger Thickness (English) | 6.2 mil |
| Finger Thickness (Metric) | 0.16 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 4.7 mil |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

