Learn More
Ansell™ TRELLCHEM™ VPS-Flash Coveralls
Catalog No. 19010014 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-014 X-Small
19-010-012 Small
19-010-011 Medium
19-010-010 Large
19-010-013 X-Large
19-010-015 2X-Large
19-010-016 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-010-014 Supplier Ansell™ Supplier No. 476520090
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Protects against hazardous chemicals in liquid, vapor, gaseous and solid form, including warfare agents
  • Made of combination of elastomers and plastics with a No-mex™ fabric
  • Chemical, flame and abrasion resistant
  • Available in both encapsulating and non-encapsulating designs
  • Option of two visors; CV or the larger VP1 made of PVC
  • Gastight chloroprene rubber coated zipper
  • Trellchem™ regulating valve has 3 ventilation rates (2, 30 and 100 l/min) plus zero/off position 
  • Glove attachments: Inner Ansell Barrier and Trellchem™ Viton™ /Butyl rubber glove
  • AlphaTec overglove for extra cut protection
  • Glove are attached with Trellchem™ Bayonet glove ring system
  • Equipped with sewn-on sock/bootie and alternatively suit has attached boots 
  • Stitched with aramid thread, taped with rubber strip on outside and barrier laminate strip welded inside
Compliance
Meets NFPA 1991 Standard on Vapor Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incident.
TRUSTED_SUSTAINABILITY
Specifications
Color Orange, Black
Material Nomex™
Size X-Small