Learn More
Ansell™ 56-903 Vinyl Closed-Back Aprons, Waist ties
Catalog No. 17987114B Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-987-114B Medium
17-987-114D X-Large
17-987-114C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-987-114B Supplier Ansell™ Supplier No. 950170
Add to Cart
Add to Cart
This item is not returnable. View return policy
With heat-sealed elastic cuffs
  • Neck ties, 15 in. long (38cm); waist ties, 29 in. long (74cm)
  • 8mil thick aprons have electronically welded vinyl binding strip that covers all edges for smoother finish
  • 6mil thick aprons have electronically welded binding strip around neck edge only; other edges are unhemmed
  • Available with snap-on nylon ties
  • Blue
Order Info
Can be cleaned to ISO Class 5 (Class 100) cleanroom standards and shipped in heat-sealed bags; call your Fisher Safety Customer Service Representative at 1-800-772-6733 for details.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Vinyl
Color Blue
Style 56-903
Thickness 6 mil
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.