Learn More
Ansell™ Vinyl Closed-Back Aprons, Plain edge
Catalog No. 19084874 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
19-084874 Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-084874 Supplier Ansell™ Supplier No. 950169
  • $870.76
    $381.65 / Case of 12
    Save $489.11
    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
With heat-sealed elastic cuffs
  • Neck ties, 15 in. long (38cm); waist ties, 29 in. long (74cm)
  • 8mil thick aprons have electronically welded vinyl binding strip that covers all edges for smoother finish
  • 6mil thick aprons have electronically welded binding strip around neck edge only; other edges are unhemmed
  • Available with snap-on nylon ties
  • Blue
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Vinyl
Color Blue
Cuff Style Elastic
Thickness 6 mil
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.