Learn More
Ansell™ Vinyl Coat Aprons
Catalog No. 19166358 Shop All Ansell Products
Change view
Click to view available options
8mil; Blue; 2X-Large
Catalog No.
19-166-358 8mil; Blue; 2X-Large
1 options
This item is not returnable. View return policy
The elastic band on this product may contain natural rubber latex which may cause allergic reactions in some individuals.
Catalog No. 19-166-358 Supplier Ansell™ Supplier No. 56 910XXL950194
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
The elastic band on this product may contain natural rubber latex which may cause allergic reactions in some individuals.
Greater body coverage with comfortable wear
  • Virgin vinyl, 8 mil thickness
  • Open back and sleeves
  • No exposed elastic or metal parts to create static and corrosion
  • Permits cleaning to Class 100 standards
  • Completely heat sealed and individually bagged
  • Medical Laboratories
  • Research Laboratories
  • Cleanrooms
  • Pharmaceutical
  • Chemical processing, oil refining and petrochemicals
  • Janitorial and maintenance work
  • Beef/pork/poultry/fish and other food processing
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Vinyl
Color Blue
Thickness 8 mil