Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 23-193 Heavy Duty Foam-Insulated PVC-Coated Gloves

Catalog No. 19019290 Shop All Ansell Products
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-019290 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-019290 Supplier Ansell Supplier No. 205059

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PVC coating remains flexible, and helps users maintain a secure grip in even the lowest temperatures.

  • PVC vinyl coating protects hands from abrasions, while also remaining soft and flexible during cold applications
  • Coating repels water, oil, and grease
  • Foam insulation helps keep hands warm in low temperatures
  • Enhanced grip for safe handling of heavy duty objects
  • Curved fingers, wing thumb, and soft jersey cotton lining
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Color Orange
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material PVC
Size 10
EN388 Standard 3121B
For Use With (Application) Metal Fabrication
Glove Cuff Style Safety
Liner Material Cotton Jersey
Series 23-193
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.